missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG(H+L) Donkey anti-Goat, Biotin, Secondary Antibody, Proteintech
Donkey anti-Goat Affinipure Secondary Antibody
50.00 € - 179.00 €
Specifications
Antigen | IgG(H+L) |
---|---|
Applications | ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry |
Conjugate | Biotin |
Form | Liquid |
Host Species | Donkey |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17630347
|
Proteintech
SA00004-3-200UL |
200 μL |
50.00 €
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17620347
|
Proteintech
SA00004-3-1ML |
1 mL |
179.00 €
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
IgG(H+L) | |
Biotin | |
Donkey | |
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. |
ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry | |
Liquid | |
Goat | |
Secondary | |
Store at -20°C. Stable for one year after shipment. |