missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ DISPAX-REACTOR™ DR
Use this high shear, three stage dispersing machine for the production of micro-emulsions and very fine suspensions. IKA™ DISPAX-REACTOR™ DR features three rotor-stator combinations (generators) in a series which produce a small droplet or particle size, with a very narrow distribution.
Brand: IKA™ U078352
This item is not returnable.
View return policy
Description
The DR line offers the same advantages as the UTL for scalability and sanitary design, and is CIP and SIP capable.
Generators can be easily interchanged, offering the ultimate in flexibility.
Applications: Generators can be easily interchanged, offering the ultimate in flexibility.
- Neutralization & PH adjustment
- Tooth paste
- Salt solutions
- Edible oil refining & degumming
- Shampoos & Conditioners
- Polymerizations
- Pesticides, Herbicides
- Reaction Enhancements
- Microencapsulations
- Viscosity Control
- Creams, Lotions
- Fuels
Specifications
Sonicator | |
Manufacturing of Fine Emulsions and Suspensions | |
Module DISPAX-REACTOR™ DR | |
1 Ea. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction