missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-10 Human ELISA Kit

Measure levels of human interleukin-10 in human serum, plasma (EDTA, heparin, sodium citrate), urine and culture supernatant samples using 96-well plates.

718.00 € - 2235.00 €

Specifications

Accession Number P22301
Assay Range 15.36-600 pg/mL
Assay Sensitivity <3 pg/mL
Description Human IL-10 ELISA Kit
Conjugate Biotin
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10443425
View Documents
Invitrogen™
EHIL102
2 x 96 Tests
770.00 €
1 set
In Stock
to see stock.
Add to basket
 
10618684
View Documents
Invitrogen™
EHIL105
5 x 96 Tests
2235.00 €
1 set
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
 
15647948
View Documents
Invitrogen™
EHIL10
96 Tests
718.00 €
Pack of 96
In Stock
to see stock.
Add to basket
 
Description

Description

Assay IL-10 (human interleukin-10) levels in human serum, plasma (EDTA, heparin, sodium citrate), urine and culture supernatant samples with our Invitrgen™ Human IL-10 ELISA Kits.

These human IL-10 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-10 (IL-10) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant human interleukin-10 (IL-10)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 15.4 to 600pg/mL

  • Sensitivity: < 3pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with human IL-1α, IL-2, IL-3, IL-4, IL-6, IL-7, IL-8, TNFα, TNFβ or IFNγ. Approx. 2% cross-reactivity with recombinant mouse IL-10

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

Specifications

Specifications

P22301
<3 pg/mL
Biotin
ELISA Kit
No significant cross-reaction with human IL-1a, IL-2, IL-3, IL-4, IL-6, IL-7, IL-8, TNFa, TNFß or IFNg. Approx. 2% cross-reactivity with recombinant mouse IL-10
Colorimetric Microplate Reader
CSIF,GVHDS,IL-10A,TGIF,IL10
<10%
Absorbance at 450nm minus absorbance at 550nm
RUO
2°C to 8°C
1 hr. 20 min.
15.36-600 pg/mL
Human IL-10 ELISA Kit
Colorimetric ELISA (TMB substrate) with Plates
Plasma, Serum, Supernatant, Urine
Human
3586
<10%
HRP
Intra-assay CV <10%; inter-assay CV <10%
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL; Urine,50 μL
Human
3 hr.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.