Learn More
Invitrogen™ IL-2 Human ELISA Kit
PD-L1 Human ELISA Kit
Brand:  Invitrogen™ BMS2212
Product Code. 15551727
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
PD-L1 Human ELISA Kit for ELISA
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P60568
9.1 pg/mL
ELISA Kit
Human
Colorimetric Microplate Reader
IL-2,TCGF,lymphokine
7%
HRP
RUO
2°C to 8°C
1 hr. 20 min.
18.8-1,200 pg/mL
Biotin
Plasma, Serum, Supernatant
ELISA
3558
5%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Controls, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL
Human
3 hr. 30 min.