missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-2 ProQuantum Immunoassay Kit

ELISA
788.00 € - 3530.00 €
Specifications
Accession Number | P60568 |
---|---|
Assay Range | 0.0128 to 5000 pg/mL |
Assay Sensitivity | <0.01 pg/mL |
Conjugate | FAM™ |
Detection Method | Taqman™ Real-time PCR |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15970483
![]() |
Invitrogen™
A35603 |
96 Tests |
788.00 €
96 reactions |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||||
17143119
![]() |
Invitrogen™
A356035 |
5 x 96 Tests |
3530.00 €
Each |
Estimated Shipment: 11-04-2025
Log in to see stock. |
|||||
Description
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.Specifications
P60568 | |
<0.01 pg/mL | |
Taqman™ Real-time PCR | |
ProQuantum | |
Human | |
qPCR Instruments | |
IL-2; TCGF; lymphokine | |
Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase | |
RUO | |
Frozen at -20°C | |
30 mins |
0.0128 to 5000 pg/mL | |
FAM™ | |
Proximity-based Immunoassay | |
Plasma, Serum, Supernatant | |
Protein Quantitation | |
3558 | |
1 hr. | |
FAM™ | |
Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL | |
IL-2 | |
2 hrs |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title