missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-2 Monoclonal Antibody (JES6-5H4), PE-eFluor™ 610, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
188.00 € - 530.00 €
Specifications
Antigen | IL-2 |
---|---|
Clone | JES6-5H4 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16676464
|
Affymetrix eBioscience
61-7021-80 |
25 μg | N/A | ||||||
16686464
|
Invitrogen
61-7021-82 |
100 μg |
530.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The JES6-5H4 antibody reacts with mouse interleukin-2 (IL-2), a 17 kDa T cell growth factor and a major immunoregulatory cytokine.
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an μgcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.Specifications
IL-2 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
16183 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IL2 |
JES6-5H4 | |
Flow Cytometry | |
PE-eFluor 610 | |
Rat | |
Mouse | |
P04351 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IL-2 Monoclonal Antibody (JES6-5H4), PE-eFluor™ 610, eBioscience™, Invitrogen™