missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IL-4 ProQuantum Immunoassay Kit

ELISA
788.00 € - 3530.00 €
Specifications
Accession Number | P07750 |
---|---|
Assay Range | 0.32 to 25,000 pg/mL |
Assay Sensitivity | <0.3 pg/mL |
Conjugate | FAM™ |
Detection Method | Taqman™ Real-time PCR |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16331994
![]() |
Invitrogen™
A42893 |
96 Tests |
788.00 €
Pack of 96 |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
17164719
![]() |
Invitrogen™
A428935 |
5 x 96 Tests |
3530.00 €
Each |
Estimated Shipment: 11-04-2025
Log in to see stock. |
|||||
Description
Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.Specifications
P07750 | |
<0.3 pg/mL | |
Taqman™ Real-time PCR | |
ProQuantum | |
Mouse | |
qPCR Instruments | |
1 hr. | |
FAM™ | |
Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL, Supernatant 2-5 μL | |
IL-4 | |
2 hrs |
0.32 to 25,000 pg/mL | |
FAM™ | |
Proximity-based Immunoassay | |
Plasma, Serum, Supernatant, Supernatant | |
Protein Quantitation | |
16189 | |
Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase | |
RUO | |
Frozen at -20°C | |
30 mins |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title