missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GenomiPhi V2 DNA Amplification Kit
Amplify whole genomes with high efficiency and representation. Cytiva Life Sciences™ illustra™ GenomiPhi V2 DNA Amplification Kit provides whole genome amplification with yields of 4 to 7μg.
224.98 € - 2982.00 €
Specifications
Content And Storage | Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C. Thaw components on ice and maintain at 0°C to 4°C during handling. |
---|---|
Description | Illustra GenomiPhi V2 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 4 to 7 μg yield from nanogram amounts of DNA sample. |
For Use With (Application) | Genomic DNA Amplification |
Product Type | illustra GenomiPhi V2 DNA Amplification Kit |
Size | > 10 kb |
Includes: Sample buffer, Reaction buffer, Enzyme mix, and Lambda control DNA (10ng/μL)
Product Code | Brand | Includes | No. of Reactions | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | No. of Reactions | Price | Quantity & Availability | ||||
10471645
|
Cytiva
25-6600-30 |
225μL Sample Buffer, 225μL Reaction Buffer, 25μL Enzyme mix, 20μL Control DNA (lambda) | 25 |
224.98 €
25 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10628505
|
Cytiva
25-6600-31 |
0.9mL Sample Buffer, 0.9mL Reaction Buffer, 100μL Enzyme mix, 20μL Control DNA (lambda) | 100 |
611.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10309494
|
Cytiva
25-6600-32 |
5 x 0.9mL Sample Buffer, 5 x 0.9mL Reaction Buffer, 5 x 100μL Enzyme mix, 20μL Control DNA (lambda) | 500 |
2982.00 €
500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Starting material can be purified DNA or nonpurified cell lysates
- Simple protocol provides typical yields of 4 to 7μg DNA and average product lengths >10kb
- Less hands-on time compared to traditional isolation methods
- Representative amplification of the whole genome
- Automation-friendly protocol with great reproducibility
- High-quality DNA for array CGH, high-throughput genotyping, DNA archival, PCR, restriction enzyme digestion, hybridization, and cloning
- Contains all of the components necessary for genomic DNA preparation by isothermal strand displacement amplification
Specifications
Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C. Thaw components on ice and maintain at 0°C to 4°C during handling. | |
Genomic DNA Amplification | |
> 10 kb |
Illustra GenomiPhi V2 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 4 to 7 μg yield from nanogram amounts of DNA sample. | |
illustra GenomiPhi V2 DNA Amplification Kit | |
1.5-2 h |
Safety and Handling
Product Identifier
Hazard Statement
Supplemental information
- GenomiPhi v2 DNA Amplification Kit, 25 reactions
Hazard Statement
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva GenomiPhi V2 DNA Amplification Kit