missing translation for 'onlineSavingsMsg'
Learn More

Cytiva NAP™ Columns, NAP-10

Use for rapid and efficient purification of DNA and oligonucleotides (>10-mer) in less than 15 minutes by gravity flow. Cytiva Life Sciences™ illustra™ NAP™ Columns are disposable columns prepacked with Sephadex™ G-25 DNA Grade

217.00 € - 497.00 €

Specifications

For Use With (Application) Rapid and efficient purification of DNA and Oligonucleotides
Includes Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution
Product Type NAP-10
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10228284
View Documents
Cytiva
17-0854-01
20 Pack
217.00 €
Pack of 20
Please to purchase this item. Need a web account? Register with us today!
10208564
View Documents
Cytiva
17-0854-02
50 Pack
497.00 €
Pack of 50
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Disposable columns prepacked with Sephadex™ G-25 DNA Grade Gel require only gravity to run
  • Available in three sizes depending on sample volume: 0.5mL (NAP-5), 1mL (NAP-10) or 2.5mL (NAP-25)
  • Useful for small-scale purification, desalting, and buffer exchange

Specifications

Specifications

Rapid and efficient purification of DNA and Oligonucleotides
Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution
NAP-10
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 5-chloro-2-methyl-2H-isothiazol-3-one, 2-methyl-2H-isothiazol-3-one

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.