Learn More
Cytiva NAP™ Columns, NAP-25
Use for rapid and efficient purification of DNA and oligonucleotides (>10-mer) in less than 15 minutes by gravity flow. Cytiva Life Sciences™ illustra™ NAP™ Columns are disposable columns prepacked with Sephadex™ G-25 DNA Grade
294.00 € - 630.00 €
Specifications
For Use With (Application) | Rapid and efficient purification of DNA and Oligonucleotides |
---|---|
Includes | Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution |
Product Type | NAP-25 |
Includes: Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing, and sample elution
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10004064
|
Cytiva
17-0852-01 |
20 Pack |
294.00 €
Pack of 20 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10186464
|
Cytiva
17-0852-02 |
50 Pack |
630.00 €
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Disposable columns prepacked with Sephadex™ G-25 DNA Grade Gel require only gravity to run
- Available in three sizes depending on sample volume: 0.5mL (NAP-5), 1mL (NAP-10) or 2.5mL (NAP-25)
- Useful for small-scale purification, desalting, and buffer exchange
Order Info
Sephadex G-25 DNA grade gel comes in a column buffer consisting of distilled water with 0.15% Kathon™ CG/ICP Biocide.
Specifications
Rapid and efficient purification of DNA and Oligonucleotides | |
Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution | |
NAP-25 |
Safety and Handling
- ||Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : reaction mass of:5-chloro2-methyl-4-isothiazolin-3-one¢EC no. 247-500-7! and 2-methyl-2H-isothiazol-
Your input is important to us. Please complete this form to provide feedback related to the content on this product.