missing translation for 'onlineSavingsMsg'
Learn More

Cytiva NAP™ Columns, NAP-5

Use for rapid and efficient purification of DNA and oligonucleotides (>10-mer) in less than 15 minutes by gravity flow. Cytiva Life Sciences™ illustra™ NAP™ Columns are disposable columns prepacked with Sephadex™ G-25 DNA Grade

201.00 € - 466.00 €

Specifications

For Use With (Application) Rapid and efficient purification of DNA and Oligonucleotides
Includes Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution
Product Type NAP-5
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10054394
View Documents
Cytiva
17-0853-01
20 Pack
201.00 €
Pack of 20
Please to purchase this item. Need a web account? Register with us today!
10218284
View Documents
Cytiva
17-0853-02
50 Pack
466.00 €
Pack of 50
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Disposable columns prepacked with Sephadex™ G-25 DNA Grade Gel require only gravity to run
  • Available in three sizes depending on sample volume: 0.5mL (NAP-5), 1mL (NAP-10) or 2.5mL (NAP-25)
  • Useful for small-scale purification, desalting, and buffer exchange

Specifications

Specifications

Rapid and efficient purification of DNA and Oligonucleotides
Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution
NAP-5
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 5-chloro-2-methyl-4-isothiazolin-3-one,2-methyl-2H-isothiazol-3-one

SDS
Documents

Documents

Product Certifications