Learn More
Cytiva PuReTaq Ready-To-Go™ PCR Beads
Pre-mixed, pre-dispensed reaction beads optimized for performing standard PCR amplifications. Cytiva Lifescience™ illustra™ PuReTaq Ready-To-Go™ PCR Beads are stable at ambient temperature.
Brand:  Cytiva 27-9558-01
 View more versions of this product
Product Code. 10289034
  • 202.00 € / 100 reactions
Explore more special offers
Description
Description
  • puReTaq DNA polymerase and high-purity reagents provide robust performance with very low levels of contaminating DNA
  • Manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
  • Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage
  • Verified for real-time PCR
  • Ready-To-Go means simply add template DNA and primers and cycle
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Room temperature stable beads containing ∼ 2.5 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer, When a bead is reconstituted to a 25μL final volume, the concentration of each dNTP
1 Kit
100
0.5mL Tube
Standard PCR
500μL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • PuReTaq
Hazard Statement
  • EUH210-Safety data sheet available on request.
Supplemental information
  • UNKNOWN ACUTE-73.3 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 73.3% of components with unknown hazards to the aquatic environment.