missing translation for 'onlineSavingsMsg'
Learn More

Cytiva RAPD Analysis Beads

For performing DNA-profiling experiments using the randomly amplified polymorphic DNA (RAPD) technique

926.00 € - 1377.00 €

Specifications

Format Tube
pH 8.3
For Use With (Application) Standard PCR
Volume 500 μL
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10409734
View Documents
Cytiva
27-9502-01
100 Reactions, 6 Primers
1377.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
10297884
View Documents
Cytiva
27-9500-01
100 Reactions
926.00 €
100 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Preformulated, single-dose, ambient-temperature-stable reaction beads ensure greater reproducibility between reactions, minimize pipetting steps and reduce the potential for pipetting errors and contamination
  • RAPD reactions are simple to perform—simply add genomic DNA solution and primer to a tube of RAPD Analysis Beads to a final reaction volume of 25 μL and cycle the reaction
  • Each lot of RAPD Analysis Beads is function-tested to ensure its ability to generate a differential banding pattern between the two control E. coli strains using RAPD Analysis Primer 2 included with the RAPD beads
  • RAPD reactions are preoptimized for use with a wide variety of organisms
  • Beads are provided in thin-walled 0.5 mL tubes compatible with most thermal cyclers
Specifications

Specifications

Tube
8.3
Standard PCR
500 μL
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.