Learn More
Cytiva RNAspin Mini Isolation Kit
Extract high-quality RNA from a wide range of samples rapidly. Cytiva Life Sciences™illustra™ RNAspin Mini Isolation Kit can be used to isolate total RNA from cultured cells, tissue, bacteria, or yeast.
Brand:  Cytiva 25-0500-71
 View more versions of this product
Product Code. 11545065
  • 311.00 € / 50 reactions
Explore more special offers
Includes: Protocol booklet with detachable, quick reference card, necessary components for specified number of preps, including prefilters and DNase I
Description
Description
  • Recovers total RNA due to removal of genomic DNA through on-column DNase I treatment
  • Can be used on cultured cells, tissue, yeast and hard-to-lyse bacterial strains
  • Prefilters reduce lysate viscosity for maximum yield and purity
  • Results can be obtained with small amounts of sample
  • Binding capacity: 100μg
  • Sample size: 30mg (animal tissue), up to 5 × 106 cells (cell culture)
  • Elution volume: 40 to 120μL
  • RNA Purity (A260/A280): 1.8 to 2.2
  • Total time of preparation: <30 min/6 preps
Quantitative reverse transcriptase PCR (QRT-PCR), Northern blot, and microarray experiments
TRUSTED_SUSTAINABILITY
Specifications
Specifications
illustra RNAspin Mini RNA Isolation Kit is designed for rapid extraction of high-quality RNA from a wide range of sample types.
Protocol booklet with detachable, quick-reference guide and necessary components, including prefilters and DNase I
illustra RNAspin Mini Kit
<30 mg
Up to 100μg
Used for the isolation of high-quality total RNA from a wide range of source material including hard-to-lyse bacterial strains. Purified RNA is suitable for use in sensitive downstream applications such as quantitative reverse transcriptase PCR (qRT-PCR),
50 Preps
1.8-2.2
< 30 min/6 preps
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501-Dispose of contents/container to .