missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephadex™ G-50 and G-100 DNA Grade Agarose Gels

Used for purification of DNA from small molecules. Cytiva illustra™ Sephadex™ G-50 and G-100 DNA Grade Agarose Gels purify DNA by gel filtration.

Brand:  Cytiva 17-0573-02

383.80 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.


 View more versions of this product

Product Code. 11584925

  • 418.00 € / 100g
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Tested to ensure reproducibly high recovery of DNA
  • Suitable for researchers who prefer to prepare their own columns for purifying nucleic acids

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 g
Auto Seq G-50 column, ProbeQuant G-50 micro column, MicroSpin G-50 column and NICK column
G-50 and G-100 DNA Grade Agarose Gels
For purification of DNA from small molecules by gel filtration
Cross-linked Dextran with Epichlorohydrin
4° to 30°C
50 to 150 μm
9 to 11 mL/g in Distilled Water
Beaded Gel
2 to 10
DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers