missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Shrimp Alkaline Phosphatase

Catalyzes removal of 5'- and 3'-phosphate groups from DNA, RNA, nucleotides, and proteins

Brand:  Cytiva E70092Y

116.14 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.


 View more versions of this product

Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10723597

  • 134.17 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 10x reaction buffer: 200mM Tris-HCl, pH 8.0, 100mM MgCl2; Dilution buffer: 50mM Tris-HCl, pH 8.0
This item is not returnable. View return policy

Description

Description

  • Bacterial
  • Expressed in E.Coli
  • Inactivated by heating at 75°C for 5 minutes
  • Active in PCR buffers and restriction endonuclease buffers
  • Activity: 1U/μL
  • Purified to homogeneity
  • Free of all contaminating endonucleases, exonucleases and ribonucleases
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Purified to homogeneity
Bacteria
500 U
Alkaline Phosphatase
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.