Learn More
Cytiva Tissue and Cells genomicPrep Mini Spin Kit
Designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from a variety of animal tissues and mammalian cell cultures
Brand:  Cytiva 28-9042-75
 View more versions of this product
Product Code. 10685545
  • 249.00 € / 50 reactions
Explore more special offers
Includes: Spin columns prepacked with a novel silica-membrane; Lyophilized proteinase K powder; Lysis solutions, wash and elution buffers; 2mL Microcentrifuge collection tubes; Full protocol booklet with a detachable, quick reference protocol card
Description
Description
To minimize the incidence of DNA shearing during extraction, the illustra tissue and cells genomicPrep Mini Spin Kit includes a lysis solution with proteinase K to gently release genomic DNA from tissues or cells. The kit can be used to extract DNA from diverse tissue types such as liver, kidney, and mouse tails with yields ranging from 0.5 to 1.5μg of genomic DNA per mg of tissue. The kit consistently generates purified genomic DNA with optical density A260/280 readings of about 1.8.
  • Capable of purifying up to 1.5μg/mg of tissue from an input tissue sample range of 5 to 50mg, and up to 40μg of genomic DNA per 5 million cells depending on cell type
  • Total DNA extraction time reduced significantly to 90 minutes (including tissue disruption)
  • Produces high-quality genomic DNA with high molecular weight and less degradation
  • Validated in several downstream applications including real-time PCR, endpoint PCR, multiplex PCR, and restriction enzyme digests
TRUSTED_SUSTAINABILITY
Specifications
Specifications
illustra tissue and cells genomicPrep Mini Spin Kit is designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from a variety of animal tissues and mammalian cell cultures in just 90 min for tissues and 45 min for ce
Spin columns prepacked with a novel Silica membrane, Lyophilized Proteinase K powder, Lysis solution, wash and Elution Buffers, microcentrifuge collection tubes, a full protocol booklet and a detachable, quick reference protocol card
illustra tissue and cells genomicPrep Mini Spin Kit
5-50 mg
Used for several downstream applications including—restriction enzyme analysis, ligation, cloning, DNA sequencing and PCR
50 Preps
1.75
90 min[Animal Tissue], 45 min[Cultured cells]
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1