missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Image-iT™ Green Hypoxia Reagent
Image-iT Green Hypoxia Reagent is a novel, fixable fluorogenic compound for measuring hypoxia in live cells.
141.00 € - 508.00 €
Specifications
Content And Storage | Store at -5 to -30°C. Protect from light. |
---|---|
Product Type | Reagent |
Product Line | Image-iT™ |
Reagent Type | Hypoxia Reagent |
Shipping Condition | Approved for shipment on Wet or Dry Ice |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15834154
|
Invitrogen™
I14834 |
1 Vial |
141.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15940773
|
Invitrogen™
I14833 |
5 Vials |
508.00 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Image-iT Green Hypoxia Reagent is a novel, fixable fluorogenic compound for measuring hypoxia in live cells. It is non-fluorescent when live cells are in an environment with normal oxygen concentrations and becomes fluorescent when oxygen levels are decreased. Image-iT Green Hypoxia Reagent sustains its fluorescence when cells/tissue return to normal oxygen levels, allowing the cells/tissue to be fixed with minimal loss of fluorescent signal.Features of Image-iT Green Hypoxia Reagent include:
- Indicates hypoxia in live cells by fluorescing in low oxygen environments
- Fixable—sustains fluorescent signal when cells/tissue return to normal oxygen levels
- Easy to use—just add to cell culture media and image
Order Info
- Store at -5 to -30°C. Protect from light.
- Stable for at least 6 months after receiving.
Specifications
Store at -5 to -30°C. Protect from light. |
|
Image-iT™ | |
Approved for shipment on Wet or Dry Ice |
Reagent | |
Hypoxia Reagent |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title