missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Image-iT™ Plant Tissue Clearing Reagent

Image-iT Plant Tissue Clearing Reagent is used to render whole-plant tissue optically clear for imaging. Image-iT Plant Tissue Hard-Set Mountant can then be used to mount and preserve the optically cleared whole-plant tissue for imaging, storage, and archiving.

Brand:  Invitrogen™ V11328

160.36 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 16399573

  • 211.00 € / 30mL
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

These reagents are compatible with both fluorescence-based and colorimetric-based applications. Image-iT Plant Tissue Clearing Reagent is an effective replacement for chloral hydrate, traditionally used to clear plant tissues in botanical microscopy.

Features of Image-iT Plant Tissue Clearing Reagent include:
• Clears plant tissue for whole-specimen imaging
• Soft-set mountant for imaging with refractive index of 1.45
• Effective replacement for chloral hydrate

Plant tissues often have structures with different refractive indexes, causing light scattering and spherical aberration, resulting in opaqueness. Image-iT Plant Tissue Clearing Reagent, with a high refractive index of 1.45, allows light to pass through the plant tissues with minimal light scattering, resulting in optical clarity and the ability to image deeper within the tissue.

Traditionally, researchers used chloral hydrate to clear plant specimens, but under current regulations it is a Federally Regulated Schedule IV substance (1), making it harder to acquire and use. Image-iT Plant Tissue Clearing Reagent is an effective replacement for chloral hydrate in almost all applications and tissue types where optical clearing is required.

Image-iT Plant Tissue Clearing Reagent and Image-iT Plant Tissue Hard-Set Mountant are based on Visikol™ for Plant Biology™ and Visikol™ Mount. Visikol for Plant Biology has been used in various fluorescent and colorimetric studies with plants (2,3,4) and has proved an effective substitute for chloral hydrate (1,5).

References:
1. Villani TS, Koroch AR, Simon JE. An improved clearing and mounting solution to replace chloral hydrate in microscopic applications. Appl Plant Sci. 2013 1(5).
2. Aoki Y, Van Trung N, Suzuki S. Impact of Piper betel leaf extract on grape downy mildew: effects of combining 4-allylpyrocatechol with eugenol, α-pinene or β-pinene. Plant Protect Sci. 2018 55: 23-30.
3. Lee KJI, Calder GM, Hindle CR, Newman JL, Robinson SN, Avondo JJHY, Coen ES. Macro optical projection tomography for large scale 3D imaging of plant structures and gene activity. J Exp Bot. 2017 68(3):527-538.
4. Vergara-Díaz O, Chairi F, Vicente R, Fernandez-Gallego JA, Nieto-Taladriz MT, Aparicio N, Kefauver SC, Araus JL. Leaf dorsoventrality as a paramount factor determining spectral performance in field-grown wheat under contrasting water regimes. J Exp Bot. 2018 69(12):3081-3094.
5. Urban MA, Barclay RS, Sivaguru M, Punyasena SW. Cuticle and subsurface ornamentation of intact plant leaf epidermis under confocal and superresolution microscopy. Microsc Res Tech. 2018 81(2):129-140.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

30 mL
Image-iT™
30 mL
Room Temperature
Store at 15°C to 25°C. Do not freeze.
When stored as directed, product is stable for (at least 6 months after receipt.

)
Tissue Clearing Reagent
Plant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.