Learn More
Invitrogen™ Image-iT™ Plant Tissue Hard-Set Mountant
Image-iT Plant Tissue Hard-Set Mountant is used for imaging, storage, and archiving of plant tissues optically cleared with Image-iT Plant Tissue Clearing Reagent.
Brand:  Invitrogen™ V11330
 View more versions of this product
Product Code. 16319583
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
These reagents are compatible with both fluorescence-based and colorimetric-based applications. Image-iT Plant Tissue mountant is based on Visikol™ Mount, which has a history of use in fluorescent and colorimetric studies with plants.
• Clears and mounts plant tissue for whole specimen imaging
• Hard-set mountant with refractive index of 1.415
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 mL
Store at 15°C to 25°C. Do not freeze.
When stored as directed, product is stable for (at least 6 months after receipt.
)
Image-iT™
Room Temperature