Learn More
Thermo Scientific™ IMAGEN™ Herpes Simplex Virus Positive Control Slide
Validate your test procedures with Thermo Scientific™ IMAGEN™ Herpes Simplex Virus Positive Control Slide
Brand:  Thermo Scientific™ S611030-2
Product Code. 13275319
  • 107.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Validate your test procedures with Thermo Scientific™ IMAGEN™ Herpes Simplex Virus Positive Control Slide. A Teflon-coated glass microscope slide with two separate 6mm diameter well areas containing acetone-fixed human fibroblast cells infected with either herpes simplex virus type 1 (HSV 1) or herpes simplex virus type 2 (HSV 2).
Sealed in foil pouch filled with nitrogen.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IMAGEN Herpes Simplex Virus Positive Control Slide
1 Slide