missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IMDM, powder

IMDM, powder

14.81 € - 110.00 €

Specifications

Certifications/Compliance ISO 13485 standards, FDA
Content And Storage 2 to 8°C
Form Powder
Product Type Iscove's modified Dulbecco's medium
With Additives Glutamine, Phenol Red, Sodium Pyruvate
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10184932
View Documents
Gibco™
42200014
1 L
14.81 €
1 L
Please to purchase this item. Need a web account? Register with us today!
11514496
View Documents
Gibco™
42200030
10 L
110.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Iscove's Modified Dulbecco's Medium (IMDM) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of Gibco™ IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ IMDM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ IMDM product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

IMDM contains no proteins, lipids, or growth factors. Therefore, IMDM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). IMDM uses a sodium bicarbonate buffer system (3.024 g/L), and therefore requires a 5-10% CO2 environment to maintain physiological pH.

Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation.
Specifications

Specifications

ISO 13485 standards, FDA
Powder
Glutamine, Phenol Red, Sodium Pyruvate
Mammalian cell culture
2 to 8°C
Iscove's modified Dulbecco's medium
No α±-Thioglycerol, No 2-Mercaptoethanol
4.4 to 4.8
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 36 Months

SDS
Documents

Documents

Product Certifications