missing translation for 'onlineSavingsMsg'
Learn More

Imetit dihydrobromide, 98%, Thermo Scientific Chemicals

Product Code. 15444749
Click to view available options
Quantity:
25 mg
50 mg
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

An extremely potent, high affinity agonist at H3 and H4 receptors

An extremely potent, high affinity agonist at H3 and H4 receptors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32385-58-3
Molecular Formula C6H12Br2N4S
Molecular Weight (g/mol) 332.06
MDL Number MFCD00153816
InChI Key DOBOYMKCRRLTRF-UHFFFAOYSA-N
Synonym imetit dihydrobromide, imetit hydrobromide, s-2-imidazol-4-yl ethyl isothiourea, s-2-1h-imidazol-4-yl ethyl isothiourea dihydrobromide, 2-1h-imidazol-4-yl ethyl carbamimidothioate dihydrobromide, s-2-imidazol-4-yl ethyl isothiourea dihydrobromide, imetit dihydrobromide hplc, s-2-imidazol-4-yl ethyl isothiourea 2hbr, s-2-imidazol-4-yl ethyl isothiourea dihydrochloride, 2-2-1h-imidazol-5-yl ethyl isothiourea dihydrobromide
PubChem CID 11957573
ChEBI CHEBI:64151
IUPAC Name 2-(1H-imidazol-5-yl)ethyl carbamimidothioate;dihydrobromide
SMILES Br.Br.NC(=N)SCCC1=CN=CN1

Specifications

Color White
Quantity 25 mg
Solubility Information Freely soluble in water,practically insoluble in acetone /n
Formula Weight 332.06
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Imetit dihydrobromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'