missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente™ Imhoff Sedimentation Cone

Sedimentation cone acc. to Imhoff without stopcock

Brand:  Lenz Laborglasintrumente™ 05670003

Product Code. 10603822

  • 175.00 € / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Sedimentation cone acc. to DIN 12672
  • Scale: up to 1000 mL
  • Ring mark: at 1000 mL
  • Subdivisions:
    • 0.1 mL (0-2 mL)
    • 0.5 mL (2-10 mL)
    • 1 mL (10-40 mL)
    • 2 mL (20-100 mL)
    • 50 mL (100-1000 mL)
Specifications

Specifications

1000 mL
DIN 12672
1
Borosilicate Glass
Yes
Not included
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers