missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Imidacloprid

Product Code. 17211166
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138261-41-3
Molecular Formula C9H10ClN5O2

Specifications

Quantity 250 mg
Melting Point 134.0°C to 146.0°C
Formula Weight 255.66
InChI Key YWTYJOPNNQFBPC-UHFFFAOYSA-N
SMILES [O-][N+](=O)NC1=NCCN1CC1=CC=C(Cl)N=C1
IUPAC Name 2-chloro-5-{[2-(nitroamino)-4,5-dihydro-1H-imidazol-1-yl]methyl}pyridine
Molecular Weight (g/mol) 255.66
Chemical Name or Material Imidacloprid
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Imidacloprid >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.