missing translation for 'onlineSavingsMsg'
Learn More
Imidazo[1,2-a]pyridine-6-carbaldehyde, ≥97%, Thermo Scientific™ Product Code.: 10391273

Imidazo[1,2-a]pyridine-6-carbaldehyde, ≥97%, Thermo Scientific™

Product Code. 10391273
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10391273

Brand: Thermo Scientific Maybridge CC33104CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 116355-16-9
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.149
MDL Number MFCD07772808
InChI Key VDWYLKPRUHCLAW-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-6-carbaldehyde, imidazo 1,2-a pyridine-6-carboxaldehyde, 6-formylimidazo 1,2-a pyridine, pubchem23883, acmc-20a31e, 6-imidazo 1,2-a pyridinecarbaldehyde, 6-imidazo 1,2-a-pyridinecarbaldehyde, h-imidazo 1,2-a pyridine-6-carbaldehyde, 4-hydroimidazo 1,2-a pyridine-6-carbaldehyde
PubChem CID 7164582
IUPAC Name imidazo[1,2-a]pyridine-6-carbaldehyde
SMILES C1=CC2=NC=CN2C=C1C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 146.15
Percent Purity 95%
Chemical Name or Material Imidazo[1, 2-a]pyridine-6-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Imidazo[1,2-a]pyridine-6-carbaldehyde, ≥97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.