missing translation for 'onlineSavingsMsg'
Learn More
Imidazo[1,2-a]pyridine-6-carboxylic acid, 95%, Thermo Scientific Chemicals Product Code.: 15425927

Imidazo[1,2-a]pyridine-6-carboxylic acid, 95%, Thermo Scientific Chemicals

Product Code. 15425927
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15425927

Brand: Thermo Scientific Alfa Aesar H33128.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139022-25-6
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD07021498
InChI Key ONOJJCTXSDBVSP-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-6-carboxylic acid, h-imidazo 1,2-a pyridine-6-carboxylic acid, imidazo 1,2-a pyridine-6-carboxylicacid, 1h-imidazo 1,2-a pyridine-6-carboxylic acid, 4-hydroimidazo 1,2-a pyridine-6-carboxylic acid, pubchem23163, acmc-209cig, ksc173s9l, 6-carboxyimidazo 1,2-a pyridine, imidazol 1,2-a pyridine-6-carboxylic acid
PubChem CID 7075376
IUPAC Name imidazo[1,2-a]pyridine-6-carboxylic acid
SMILES OC(=O)C1=CN2C=CN=C2C=C1

Specifications

Melting Point 244°C to 247°C
Quantity 1 g
Formula Weight 162.15
Percent Purity 95%
Chemical Name or Material Imidazo[1,2-a]pyridine-6-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10173655 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33128.03,H33128.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Imidazo[1,2-a]pyridine-6-carboxylic acid, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.