missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-8-carboxylic acid hydrochloride, 97%, Thermo Scientific™

Product Code. 11906711
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 145335-90-6
Molecular Formula C8H6N2O2·ClH
Molecular Weight (g/mol) 198.61
InChI Key XRQZBGADESKIBB-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-8-carboxylic acid hydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid monohydrochloride, imidazo 1,2-a pyridine-8-carboxylic acid, monohydrochloride, imidazo 1,2-a pyridine-8-carboxylic acid monohydrochloride, imidazo 1,2-a pyridine-8-carboxylicacidhydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid hydrochloride, imidazo 1,2-alpha pyridine-8-carboxylic acid hydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid monohydr
PubChem CID 11715356
IUPAC Name imidazo[1,2-a]pyridine-8-carboxylic acid;hydrochloride
SMILES C1=CN2C=CN=C2C(=C1)C(=O)O.Cl

Specifications

Melting Point 298°C to 300.1°C
Color White
Elemental Analysis 46.44 to 50.32% C, 13.54 to 14.67% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 198.61
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Imidazo[1, 2-a]pyridine-8-carboxylic acid hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Imidazo¢1,2-a!pyridine-8-carboxylic acid hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'