missing translation for 'onlineSavingsMsg'
Learn More
Imidazo[2,1-b]thiazole-6-carbaldehyde 97+%, Thermo Scientific™ Product Code.: 10548063

Imidazo[2,1-b]thiazole-6-carbaldehyde 97+%, Thermo Scientific™

Product Code. 10548063
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10548063

Brand: Thermo Scientific Maybridge CC32504DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 120107-61-1
Molecular Formula C6H4N2OS
Molecular Weight (g/mol) 152.171
InChI Key VYBDPAAWKNKEKY-UHFFFAOYSA-N
Synonym imidazo 2,1-b thiazole-6-carbaldehyde, imidazo 2,1-b 1,3 thiazole-6-carbaldehyde, imidazo 2,1-b thiazole-6-carboxaldehyde, acmc-20moqu, imidazo 2,1-b 1,3-thiazoline-6-carbaldehyde, imidazo 2,1-b 1,3 thiazole-6-carboxaldehyde, imidazo 2,1-b thiazole-6-carboxaldehydeimidazoShow MoreShow Less
PubChem CID 4961258
IUPAC Name imidazo[2,1-b][1,3]thiazole-6-carbaldehyde
SMILES C1=CSC2=NC(=CN21)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 152.18
Percent Purity 97%
Chemical Name or Material Imidazo[2, 1-b]thiazole-6-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Imidazo[2,1-b]thiazole-6-carbaldehyde 97+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.