missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidazole, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
2 kg
10 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 86.0°C to 90.0°C |
Color | White to Yellow |
Boiling Point | 255.0°C to 256.0°C |
Flash Point | 145°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Plastic Bottle |
Quantity | 500 g |
Beilstein | 23, 45 |
Fieser | 01,492; 02,220 |
Show More |
Safety and Handling



Product Identifier
- Imidazole
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 C
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H360D-May damage the unborn child.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Imidazole, 99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction