Learn More
Imidazole, 99%, Thermo Scientific Chemicals
Description
Imidazole is used as a buffer in the range of pH 6.2-7.8. It is also an histamine antagonist. It acts as a chelator and forms complexes with various divalent cations. It is used as a corrosion inhibitor on certain transition metals such as copper. Its derivatives, like polybenzimidazole (PBI), act as fire retardants. It finds application in photography and electronics. Imidazole salts are used as ionic liquids and precursors to stable carbenes. Imidazole derivatives like ketoconazole, miconazole and clotrimazole are involved in the treatment of various systemic fungal infections. It is a part of the theophylline molecule, present in tea leaves and coffee beans, which stimulates the central nervous system.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Imidazole is used as a buffer in the range of pH 6.2-7.8. It is also an histamine antagonist. It acts as a chelator and forms complexes with various divalent cations. It is used as a corrosion inhibitor on certain transition metals such as copper. Its derivatives, like polybenzimidazole (PBI), act as fire retardants. It finds application in photography and electronics. Imidazole salts are used as ionic liquids and precursors to stable carbenes. Imidazole derivatives like ketoconazole, miconazole and clotrimazole are involved in the treatment of various systemic fungal infections. It is a part of the theophylline molecule, present in tea leaves and coffee beans, which stimulates the central nervous system.
Solubility
Miscible with water, ethanol, ether, acetone, chloroform, pyridine and methanol. Slightly miscible with benzene and petroleum ether.
Notes
Incompatible with acids, acid anhydrides and strong oxidizing agents.
Specifications
Melting Point | 88°C to 92°C |
Density | 1.03 |
Boiling Point | 256°C |
Flash Point | 145°C (293°F) |
Assay Percent Range | 99% |
Quantity | 100 g |
Beilstein | 103853 |
Sensitivity | Hygroscopic |
Solubility Information | Miscible with water,ethanol,ether,acetone,chloroform,pyridine and methanol. Slightly miscible with benzene and petroleum ether. |
Formula Weight | 68.08 |
Show More |
RUO – Research Use Only