missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imject™ EDC BSA Spin Kit

Easily prepare immunogens from carboxyl-containing peptide antigens with bovine serum albumin (BSA) and different conjugation chemistries.

Brand:  Thermo Scientific™ 77672

Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11804045

  • 692.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Sufficient For: 5 EDC conjugation reactions and complete immunogen preparations, each with up to 2 mg peptide
• Imject BSA (in MES buffer), 5 x 2 mg
• Imject EDC Conjugation Buffer, 30 mL
• EDC Crosslinker, 5 x 10 mg
• Imject Purification Buffer Salts, 5 x 0.84 g
• Zeba Spin Desalting Columns, 7K MWCO, 2 mL, 5 columns

Store at 4°C.
Kit
Imject™
Imject EDC BSA Spin Kit
5 x 2 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Imject™ EDC BSA Spin Kit > EDC Conjugation Kit; 5 x 2mg kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.