missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imject™ cBSA (in PBS)

Prepare hapten-carrier conjugates to generate antibodies using this convenient and highly immunogenic, cationized BSA carrier protein.

Brand:  Thermo Scientific™ 77150

Product Code. 11854071

  • 461.00 € / 10mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Imject Cationized Bovine Serum Albumin (cBSA) functions as an easy-to-use and highly immunogenic carrier protein for preparing antigens for use in antibody production.

Imject cBSA is prepared by modifying native BSA with excess ethylenediamine, essentially capping all negatively-charged carboxyl groups with positively-charged primary amines. The result is a highly positively-charged protein (pI > 11) that has significantly increased immunogenicity compared to native BSA. In addition, the increased number of primary amines provides for a greater number of antigen molecules to be conjugated with typical crosslinking methods.

Imject cBSA is an excellent choice for immunogen preparation when greater immunogenicity is needed relative to BSA but conditions are not amenable to using keyhole limpet hemocyanin (KLH) or Blue Carrier™ Protein.

Highlights:

  • Convenient – purified bovine serum albumin (BSA) that is cationized and supplied lyophilized in PBS

  • Single polypeptide – modified BSA protein with molecular mass greater than 67,000 daltons

  • Multiple conjugation sites – contains 59 lysine residues, plus numerous other primary amines added by cationization, most of which are capable of reacting with a conjugation reagent to attach antigens

  • Net positive charge – confers solubility in conditions not suitable for the negatively-charged native BSA

  • Immunogenic – significantly greater immunogenicity than native BSA because the positively charged protein is processed differently by the immune-response system

TRUSTED_SUSTAINABILITY
Specifications

Specifications

4°C
10 mg
Imject™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZE393825 Certificate TypeCertificate of Analysis Date21/05/2024 Catalog Number77150
Lot NumberZA391867 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number77150
Lot NumberYL375730 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number77150
Lot NumberYC367800 Certificate TypeCertificate of Analysis Date11/03/2023 Catalog Number77150
Lot NumberXH362767 Certificate TypeCertificate of Analysis Date13/08/2022 Catalog Number77150
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Imject™ cBSA (in PBS) > cBSA in PBS; 10mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.