Learn More
Thermo Scientific™ Imject™ Maleimide-Activated Blue Carrier™ Protein
Make hapten-carrier immunogens comprising cysteine-containing peptides and other thiolated antigens with sulfhydryl-reactive immunogenic hemocyanin (CCH).
Brand: Thermo Scientific™ 77664
View more versions of this product
Description
Thermo Scientific™ Maleimide-Activated Blue Carrier Protein is sulfhydryl-reactive immunogenic hemocyanin (CCH) for hapten-carrier conjugation with cysteine-containing peptides and other thiolated antigens.
Blue Carrier Protein is a large homocyanin that is highly immunogenic like keyhole limpet hemocyanin (KLH) yet is more soluble. The carrier is rendered sulfhydryl-reactive by chemical modification with Sulfo-SMCC crosslinker, and the resulting maleimide-activated protein is then stabilized and lyophilized in phosphate buffer. Maleimide groups form covalent crosslinks with sulfhydryl (-SH) moieties on cysteine residues of peptides and other thiol-containing haptens. Numerous antigen molecules can be conjugated to each Blue Carrier Protein molecule to make them highly immunogenic and increase the likelihood of eliciting immunized animals to produce high-titer antisera to the epitopes of interest.
Highlights:
Optimized activation – specifications require that each lot is activated with more than 400 moles of sulhydryl-reactive maleimide per mole of carrier, ensuring consistently high levels of conjugatio
Sulfhydryl-reactive – engineered for conjugating cysteine-containing peptide antigens to make effective immunogens for high-titer antibody production
Simple, rapid procedure – complete the hapten-carrier conjugation reaction in two hours to yield sufficient immunogen for several injections and booster immunizations of host animals
High-quality carrier protein – Blue Carrier Protein is ultrapure Concholepas concholepas hemocyanin (CCH) prepared from molluscan hemolymph
Highly immunogenic and soluble – CCH has a high molecular mass that elicits a stronger immune response than BSA or ovalbumin
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Upon receipt store at -20°C. Product is shipped on ice. | |
10 mg | |
Blue Carrier™, Imject™ | |
Imject Maleimide-Activated Blue Carrier Protein |