missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imject™ EDC mcKLH Spin Kit

Prepare immunogens (hapten-carrier conjugates) with your antigen of choice and mariculture KLH using different conjugation chemistries.

Brand:  Thermo Scientific™ 77671

Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11803855

  • 680.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Imject Mariculture Keyhole Limpet Hemocyanin (mcKLH) is formulated to achieve optimum performance with different conjugation chemistries, and includes a complete kit for carbodiimide-mediated hapten-carrier preparation.

Mariculture KLH (mcKLH) is purified and lyophilized in buffers that optimize its stability and solubility for hapten conjugation. The large, complex protein contains hundreds of sites per molecule for effective conjugation of peptides and other antigens using amine-reactive or carboxyl-reactive crosslinkers.

Preparations in phosphate buffer are ready for hapten-carrier conjugation via amine-reactive NHS-ester or glutaraldehyde crosslinking. Preparations in MES buffer are optimized for peptide-carrier conjugation via the carboxyl-reactive carbodiimide crosslinker called EDC (or EDAC). The Imject EDC Spin Kit includes ready-to-use units of KLH carrier protein, optimized reaction buffers and trouble-free spin desalting columns for immunogen preparation using the EDC method.

Highlights:

  • Convenient packages – choose lyophilized KLH protein or complete EDC conjugation kit

  • High-yield conjugation – each molecule of KLH contains hundreds of primary amines available for coupling haptens via EDC or NHS ester crosslinkers

  • Validated quality – purified and stabilized mariculture KLH maintains solubility in aqueous solutions, unlike traditional sources of the carrier protein

  • Sustainable source – KLH is harvested from select populations of the mollusk Megathura crenulata (keyhole limpet) that are grown in mariculture, not extracted from wild populations

  • Highly immunogenic – KLH has a high molecular mass (0.5 to 13 million daltons; aggregates of 350 and 390kDa subunits) and elicits a stronger immune response than BSA or ovalbumin

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sufficient For: 5 EDC conjugation reactions and complete immunogen preparations, each with up to 2 mg peptide
• Imject Mariculture KLH (in MES buffer), 5 x 2 mg
• Imject EDC Conjugation Buffer, 30 mL
• EDC Crosslinker, 5 x 10 mg
• Imject Purification Buffer Salts, 5 x 0.84 g
• Zeba Spin Desalting Columns, 7K MWCO, 2 mL, 5 columns

Store at 4°C.
Kit
Imject™
Imject EDC mcKLH Spin Kit
5 x 2 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.