Learn More
Hughes Immersion Heated 1500 L Tank Shower
Freeze protected emergency tank shower for use in non-hazardous environments. Designed to meet and exceed ANSI standards.
Brand:  Hughes HSHM14K1SS
Product Code. 17782348
  • 18945.00 € / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Immersion heated emergency tank shower suitable for use in non-hazardous environments
  • Designed to meet and exceed ANSI standards by providing a guaranteed flow of 76 liters per minute for 15 minutes, even when there is no mains water supply
  • Eye/face wash unit mounted internally
Specifications
Specifications
1500 L
Stainless Steel
Tank Shower
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hughes Immersion Heated 1500 L Tank Shower > Tank Shower; Capacity: 1500 L