Learn More
Hughes Immersion Heated 350 L Tank Shower
Immersion heated 350 liter emergency tank shower with galvanized mild steel framework.
Brand:  Hughes HSHM14K3SS
Product Code. 17792348
  • 10100.00 € / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Immersion heated emergency tank shower for use in nonhazardous environments
  • Suitable for use where insufficient water pressure exists
  • An integral immersion heater, controlled by a dual safety thermostat, maintains the water in the tank at a constant temperature
  • Side mounted eye/face wash unit
Specifications
Specifications
350 L
Stainless Steel
Tank Shower