missing translation for 'onlineSavingsMsg'
Learn More

Leica Microsystems Immersion Oil for Microscopes

Product Code. p-8001080
Click to view available options
Refractive Index:
Type F
Unit Size:
10mL
This item is not returnable. View return policy

Product Code. 11944399

Brand: Leica Microsystems 11513859

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Use Leica™ Immersion Oil with microscope to increase the resolving power and aperture of the objective lens.

  • 10mL bottle
  • For Leica Microscopes

Specifications

Type Immersion Oil
Packaging Type 1 Pack
Quantity 10 mL
Refractive Index Type F
Size 10mL
compliance-icons
Product Identifier
  • Type F Immersion liquid
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H315-Causes skin irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.