Learn More
Ebro Electronic™ Thermocouple Rod Probes
Rod probes for temperature measurement of air, ovens, fluid and gases.
Brand:  Ebro Electronic™ 1341-0400
 View more versions of this product
Product Code. 10289969
  • 151.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For various applications in food, laboratory, industial use.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RoHS Directive 2011/65/EU
Single-use
15 mm
300 mm
NiCrNi rod
Rod Probe
40°C
TFN 520
15 mm
1.5 mm
Temperature measurement of air, ovens, fluids and gases.
300 mm
TPN 120-30
0.12 kg
1100°C
-40° to 1100°C
DIN IEC 584, Class 2