Learn More
Cytiva Immobiline™ DryPlate for IEF
Obtain reproducible IEF separations with dry, precast gels containing immobilized pH gradients
Brand:  Cytiva 80112828
Product Code. 11723339
  • 965.00 € / Pack of 3
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Immobiline DryPlate precast gels contain preformed, immobilized, non-drifting pH gradients. The gels are rehydrated to a thickness of 0.5mm in the reswelling cassette accessory before use.
  • Medium range covering 3 pH units
  • Handle mechanically stable, plastic-backed gels with confidence
  • Immobiline technology produces an absolutely stable pH gradient, allowing extremely high voltages and high resolution over narrow pH ranges, even for low mobility proteins that require long focusing times
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IEF
Precast Gel
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Hazard Statement
  • EUH210-Safety data sheet available on request.
Supplemental information
  • UNKNOWN ACUTE-90 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 90% of components with unknown hazards to the aquatic environment.