missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Immobiline™ DryStrip Gels

Product Code. 10146414 Shop All Cytiva Products
Click to view available options
pH Range:
3 to 10
3 to 11
4 to 7
6 to 11
6 to 9
Gradient:
Linear
Non-linear
Length (Metric):
7 cm
11 cm
13 cm
18 cm
24 cm
Unit Size:
Pack of 12
This item is not returnable. View return policy

Product Code. 10146414

Brand: Cytiva 17600373

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Represent a comprehensive selection of narrow-, medium- and wide-range IPG strips. Cytiva Life Sciences™ Immobiline™ DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate application.

Choose from a comprehensive range of overlapping Immobiline DryStrip gels that cover narrow (1 pH unit), medium (3 to 5 pH units), and broad (8 pH units) pH ranges. The pH gradients at the extreme ends of the pH scale are nonlinear to distribute the proteins evenly over the gel length to obtain maximum resolution.

Five narrow pH range (1 pH unit), 24-cm IPG strips deliver high resolution and protein loading capacity, which results in well separated spots with high protein content after the second-dimension separation

  • Select the broad pH 3 to 11 Non-Linear (NL) gradient Immobiline DryStrip gel for fast and efficient screening to gain a broad overview of total protein disribution
  • For more detailed studies or when using greater protein loads, choose from four new overlapping strips covering the pH 3 to 11 range
  • Higher resolution can be obtained using narrow-range (1 pH unit) Immobiline DryStrip gels
  • Use matching IPG Buffers for optimized running conditions
  • Use IPG strips directly after reswelling without further manipulation
  • Bar coded strips for convenient sample and batch tracking

  • Immobiline DryStrip gels are rehydrated with a matching IPG Buffer prior to use
  • Samples can be included directly in the rehydration solution for efficient loading of dilute samples or for loading larger amounts of protein
  • Samples can also be loaded after rehydration

TRUSTED_SUSTAINABILITY

Specifications

Description Immobiline DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate application.
Color Colorless
pH Range 3 to 11
Product Type Precast Gel
Separation Type Isoelectric Focusing
Form Dried
For Use With (Application) Industrial applications: analytical chemistry, research
Gradient Non-linear
Storage Requirements −20°C
Thickness (English) 0.19 in.
Thickness (Metric) 0.05 cm
Length (Metric) 7 cm
Width (English) 0.118 in.
Width (Metric) 0.3 cm
Show More Show Less
Hazard Statement
  • EUH210-Safety data sheet available on request.
Supplemental information
  • UNKNOWN ACUTE-20.2 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 20.2% of components with unknown hazards to the aquatic environment.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.