Learn More
Cytiva Immobiline™ DryStrip Kit and Components
Precast Immobiline DryStrip gels for running the first dimension of a 2-D electrophoresis experiment with the Multiphor II electrophoresis system
Brand:  Cytiva 18-1004-34
 View more versions of this product
Product Code. 10239624
  • 176.00 € / Pack of 4
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Run up to 12 Immobiline DryStrip strips simultaneously with up to 100μL on each Immobiline DryStrip.
  • The instruction manual presents all necessary information for the entire method for horizontal 2-D electrophoresis. By appropriate adaptations, the SDS electrophoresis can also be performed using vertical electrophoresis equipment
  • Immobiline DryStrip kit facilitates sample application and running and equilibration of Immobiline DryStrip for the first dimension in 2-D (two-dimensional) electrophoresis
  • Isoelectric focusing with an immobilized pH gradient (IPG), makes true isoelectric focusing possible and significantly improves the reproducibility of the spot distribution along the pH gradient axis of 2-D maps
  • Individual components are also available separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Multiphor II Electrophoresis System
Dry Strip Slinger