missing translation for 'onlineSavingsMsg'
Learn More

Merck Immobilon-PSQ PVDF Membranes

Immobilon Transfer Membranes offer the convenience of pre-cut sheets with the flexibility of membrane rolls

Brand:  Merck ISEQ00005

Product Code. 15760319

  • 415.00 € / Each
Estimated Shipment: 15-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Compatible stains: Coomassie brilliant blue, Amido black, India ink, Ponceau-S red, Colloidal gold, CPTS, Toluidine blue, Transillumination, Sypro™ ruby

hydrophobic, transfer, membrane, pvdf

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Filter code: ISEQ; I mmobilon-PSQ transfer membrane
Electrostatic, hydrophobic
ISEQ
Roll
0.2 μm
Western blotting; Immunoblotting (Western); Dot blot
White
Plain
PVDF
1.875 m x 26.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers