missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Breakable Modules Clear

Choose from four surfaces and a variety of wells and configurations with these immuno breakable modules, which offer an optimal choice of products for the specific assay.

Brand:  Thermo Scientific™ 446490

390.41 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.


502.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10449121

  • 507.02 € / Pack of 60
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Choose from four surfaces and a variety of wells and configurations with Thermo Scientific™ Immuno Breakable Modules, which offer reliability, well-to-well reproducibility and an optimal choice of products for the specific assay.

Choose between LockWell, BreakApart and Breakable 1x8 modules in standard 96-well frames.

Features:

  • For solid-phase immunoassays, e.g. ELISA
  • Choice of four surfaces for optimal binding
  • Free-standing modules are easy to handle
  • Alphanumeric system for easy orientation
  • Fits standard equipment
  • High optical quality for reliable results
  • Polystyrene

Choice of Surface:

  • PolySorp™, Immulon™ 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
  • MediSorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
  • MaxiSorp™, Immulon 4 HBX, and Enhanced Binding (EB) are hydrophilic and ideal for antibody sandwich assays
  • MultiSorp™ are very hydrophilic and work well for glycans and water-soluble proteins
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Breakable Strips
No
Without Lid
Colorimetric
10 Pk.
Frame
350 μL
250 μL
Round, C Bottom
60
None
Very Hydrophilic
Strip Plate
High optical quality for reliable results
8 x 12
Clear
Polystyrene
96
MultiSorp
For glycans and water soluble proteins
10
Glycans
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number181709 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number446490
Lot Number170713 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number446490
Lot Number168503 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number446490
Lot Number177304 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number446490
Lot Number173557 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number446490
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Immuno Breakable Modules Clear > Nunc; C8 LockWell; MultiSorp; 350μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.