missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Immuno Breakable Modules in White and Black
Keep wells in place with these breakable modules in white and black, ensuring fluorescence and luminescence assays are easy to handle.
Brand: Thermo Scientific™ 463201
This item is not returnable.
View return policy
Description
Keep wells securely in place with Thermo Scientific™ Breakable Modules in White and Black, ensuring fluorescence and luminescence assays are easy to handle.
Features:
- High-quality polystyrene wells
- Choice of two passive surfaces to enable assay optimization
- C8 strips provide flexibility with sample size and optimal washing performance
- Easy to handle; single well held as easily as strips
- Compatible with common instrumentation
Choice of Surface:
- Thermo Scientific™ PolySorp™ and Universal Binding (UB) are for adsorption of hydrophobic molecules.
- Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assays.
White:
- Opaque and reflective surface
- Low cross talk and minimal background
Black:
- Opaque and light-absorbing surface
- Minimum cross talk and back-scattered light
Note:
Availability may vary by country.
Specifications
Breakable Strips | |
Yes | |
Without Lid | |
8 x 12 | |
Adsorption | |
Microplate Strip | |
350 μL | |
250 μL | |
Round, C-bottom | |
60 | |
None |
Hydrophilic | |
Strip Plate | |
Chemiluminescence | |
10 Pk., 60 Cases | |
White | |
Polystyrene | |
96 | |
MaxiSorp | |
Luminescence | |
10 | |
Antibodies (Glycoproteins) |