Learn More
Thermo Scientific™ Immuno Standard Modules White and Black
BIOPHARMA
Conduct fluorescence- and luminescence-based immunoassays and binding assays using these immuno standard modules white and black.
Brand:  Thermo Scientific™ 437915
 View more versions of this product
Product Code. 10080611
  • 413.09 € / Pack of 60
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo Scientific™ Immuno Standard Modules White and Black.
Features:
  • Available in 8-, 12- or 16-well strips
  • Framed or unframed for choice of optimal assay
  • Flat-bottom or C-shaped wells
  • Choice of three surface types for optimal binding
  • White format for highest possible signal reflection with low background fluorescence; recommended for epifluorescence
  • Black format to reduce backscatter light, which may be encountered in epifluorescence
  • Transparent or clear format for use in time-resolved fluorescence applications only
Choice of Surface:
  • Thermo Scientific™ PolySorp™ and Microlite™ 1+ are for adsorption of hydrophobic molecules.
  • Microlite 2+ is slightly hydrophilic and binds a diverse range of biomolecules.
  • Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assay.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No
Yes
Colorimetric
60 Cases
Microplate Strip
350 μL
250 μL
MaxiSorp
ELISA
5
60/Cs.
Hydrophilic
Standard
8 x 12
Clear
Polystyrene
96
Non-sterile
Round, C-bottom
60
None
Antibodies (Glycoproteins)