missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Immuno Standard Modules White and Black
Conduct fluorescence- and luminescence-based immunoassays and binding assays using these immuno standard modules white and black.
Brand: Thermo Scientific™ 475515
259.15 EUR valid until 2024-09-27
Use promo code "21650" to get your promotional price.
This item is not returnable.
View return policy
Description
Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo Scientific™ Immuno Standard Modules White and Black.
Features:
- Available in 8-, 12- or 16-well strips
- Framed or unframed for choice of optimal assay
- Flat-bottom or C-shaped wells
- Choice of three surface types for optimal binding
- White format for highest possible signal reflection with low background fluorescence; recommended for epifluorescence
- Black format to reduce backscatter light, which may be encountered in epifluorescence
- Transparent or clear format for use in time-resolved fluorescence applications only
Choice of Surface:
- Thermo Scientific™ PolySorp™ and Microlite™ 1+ are for adsorption of hydrophobic molecules.
- Microlite 2+ is slightly hydrophilic and binds a diverse range of biomolecules.
- Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assay.
Specifications
No | |
Yes | |
Fluorescence | |
60 Cases | |
Black | |
Polystyrene | |
96 | |
MaxiSorp | |
Fluorescence | |
5 | |
Antibodies (Glycoproteins) |
Hydrophilic | |
Standard | |
8 x 12 | |
Adsorption | |
Microplate Strip | |
400 μL | |
350 μL | |
Round, Flat Bottom (F-well) | |
60 | |
None |