missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Tubes and Stoppers

Product Code. 10254071
Click to view available options
Material:
Polyethylene
Polystyrene
Diameter (Metric) Outer:
11 mm
12 mm
15 mm
Product Type:
Immuno Star Tubes and Stopper
Immuno Tubes and Stopper
Stopper
For Use With (Equipment):
11 mm Nunc-Immuno Tubes
12 mm Nunc-Immuno Tubes
15 mm Nunc-Immuno Tubes
Immuno Tube
Unit Size:
Pack of 1200
Pack of 1800
Pack of 2400
Pack of 3000
Pack of 3600
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 10254071

Marca: Thermo Scientific™ 443990

Solo quedan null
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones

Reproduce results time after time with these immuno tubes and stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays.

Reproduce results time after time with Thermo Scientific™ Immuno Tubes and Stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays. These products are backed by industry-leading quality standards and certifications.

Features:

  • Offered in choice of three surfaces for optimal binding
  • Three tube sizes with stoppers available (ordered separately)
  • Fits standard equipment

Choice of Surface:

  • Thermo Scientific™ PolySorp™: a hydrophobic surface that preferentially binds biomolecules that have hydrophobic domains
  • Thermo Scientific™ MaxiSorp™: a hydrophilic surface that is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character
  • Thermo Scientific™ Mini-Sorp™: a polyethylene-based material that provides a low-binding surface

Liquid Phase Immunoassay Tubes (Cat. Nos. 443990, 468608):

  • MiniSorp surface
  • Molded from special polyethylene formulation
  • For liquid phase immunoasssays, including RIA, for storing of reagents or collecting chromatographic fractions

Solid Phase Immunoassay Tubes:

  • PolySorp and MaxiSorp surfaces available for choice of optimal binding
  • For solid phase immuno techniques such as IRMA, ELISA and ILMA
  • Certified binding homogeneity
  • High-quality polystyrene for optimal quality
  • StarTube design gives higher sensitivity and faster assay times
  • Enhances assay sensitivity and reduces incubation times

Recommended for:

  • ELISA
  • Solid Phase RIA
  • Liquid Phase RIA
  • Sample storage
TRUSTED_SUSTAINABILITY

Especificaciones

Material Polyethylene
Product Type Immuno Tubes and Stopper
Surface MiniSorp
Length (Metric) 75 mm
Diameter (Metric) Outer 12 mm
No. per Pack 100
No. per Case 3000
Volume (Metric) 5 mL
For Use With (Equipment) Immuno Tube
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado