missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Immuno Tubes and Stoppers
Click to view available options
Material:
Polyethylene
Polystyrene
Diameter (Metric) Outer:
11 mm
12 mm
15 mm
Product Type:
Immuno Star Tubes and Stopper
Immuno Tubes and Stopper
Stopper
For Use With (Equipment):
11 mm Nunc-Immuno Tubes
12 mm Nunc-Immuno Tubes
15 mm Nunc-Immuno Tubes
Immuno Tube
Unit Size:
Pack of 1200
Pack of 1800
Pack of 2400
Pack of 3000
Pack of 3600
Description
Reproduce results time after time with Thermo Scientific™ Immuno Tubes and Stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays. These products are backed by industry-leading quality standards and certifications.
Features:
- Offered in choice of three surfaces for optimal binding
- Three tube sizes with stoppers available (ordered separately)
- Fits standard equipment
Choice of Surface:
- Thermo Scientific™ PolySorp™ is a hydrophobic surface that preferentially binds biomolecules that have hydrophobic domains.
- Thermo Scientific™ MaxiSorp™ is a hydrophilic surface that is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character.
- Thermo Scientific™ Mini-Sorp™ is a polyethylene-based material that provides a low-binding surface.
Liquid Phase Immunoassay Tubes (Cat. Nos. 443990, 468608):
- MiniSorp surface
- Molded from special polyethylene formulation
- For liquid phase immunoasssays, including RIA, for storing of reagents, or collecting chromatographic fractions
Solid Phase Immunoassay Tubes:
- PolySorp and MaxiSorp surfaces available for choice of optimal binding
- For solid phase immuno techniques such as IRMA, ELISA and ILMA
- Certified binding homogeneity
- High–quality polystyrene for optimal quality
- StarTube design gives higher sensitivity and faster assay times
- Enhances assay sensitivity and reduces incubation times
Recommended for:
- ELISA
- Solid Phase RIA
- Liquid Phase RIA
- Sample storage
Specifications
Specifications
| Material | Polystyrene |
| Product Type | Immuno Tubes and Stopper |
| Surface | MaxiSorp |
| Length (Metric) | 70 mm |
| Diameter (Metric) Outer | 11 mm |
| Certifications/Compliance | MaxiSorp tubes are certified for IgG binding (tube to tube % CV less than 5%) |
| No. per Pack | 180 |
| No. per Case | 1800 |
| Volume (Metric) | 4 mL |
| For Use With (Equipment) | Immuno Tube |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction