missing translation for 'onlineSavingsMsg'
Learn More

Manutan In/out board with 20 names 360mm x 382mm

Brand:  Manutan A337603

Encompass_Preferred

Product Code. 12868139

  • 255.41 € / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Highly durable in/out board, ideally suited for use in industrial environments. Slide button design allows easy use. Supplied with card inserts. Easy fitting with fixings included.
  • 20 name tabs
  • Dimensions (H x W) 360mm x 382mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers